Ethyl 4-bromo-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

A341101-100G A341101-10G A341101-1G A341101-25G A341101-5G
103825-826EA 6 USD
103825-826 103836-804 76828-484 76828-486 76839-826 76839-828 76839-830
Ethyl 4-bromo-2-fluorobenzoate 98%
Ethyl 4-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204444
CAS Number: 474709-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR