Ethyl 4-aminothiazole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: ethyl 4-amino-1,3-thiazole-5-carboxylate

A366896-100MG A366896-250MG A366896-25G A366896-1G A366896-5G
103826-412EA 30 USD
103826-412 103837-426 76817-452 76817-454 76817-456 77306-784 77306-786
Ethyl 4-aminothiazole-5-carboxylate 95%
Ethyl 4-aminothiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
Boiling Pt: 300.7 °C (760 mm Hg)
Flash Pt: 135.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798219
CAS Number: 152300-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR