Ethyl-4,6-dichloronicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,6-dichloronicotinate

A278526-1G A278526-25G A278526-100G A278526-10G A278526-500G
103813-416EA 9 USD
103813-416 103853-358 77093-284 77093-286 77099-132 77099-134 77099-136
Ethyl-4,6-dichloronicotinate 98%
Ethyl-4,6-dichloronicotinate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
Boiling Pt: 276.3 °C (760 mm Hg)
Melting Pt: 32…34 °C
Storage Temperature: Ambient
MDL Number: MFCD00234408
CAS Number: 40296-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR