Ethyl 4,6-dichloro-2-methylnicotinate 97%

Supplier: Ambeed
Warning

A144974-10G A144974-1G A144974-5G
103803-474EA 9 USD
103803-474 103841-552 77027-834 77316-816 77316-818
Ethyl 4,6-dichloro-2-methylnicotinate 97%
Ethyl 4,6-dichloro-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD05666193
CAS Number: 686279-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR