Ethyl-4,4-difluorocyclohexanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,4-difluorocyclohexanecarboxylate

A182880-100G A182880-10G A182880-1G A182880-25G A182880-5G
103815-578EA 10 USD
103815-578 103846-140 76690-626 76690-628 77083-714 77083-716 77083-718
Ethyl-4,4-difluorocyclohexanecarboxylate 98%
Ethyl-4,4-difluorocyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄F₂O₂
MW: 192.21 g/mol
Boiling Pt: 203.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03788494
CAS Number: 178312-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR