4,4-Diethoxy-3-oxo-butyric acid ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,4-diethoxy-3-oxobutanoate

A157797-250MG A157797-1G A157797-5G
103805-046EA 126 USD
103805-046 103843-250 76812-544 77300-292 77300-294
4,4-Diethoxy-3-oxo-butyric acid ethyl ester 95%
4,4-Diethoxy-3-oxo-butyric acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₅
MW: 218.247 g/mol
Boiling Pt: 292.2±25.0 °C (760 mm Hg)
Flash Pt: 131.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22494947
CAS Number: 10495-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR