Ethyl-4,4,4-trifluorobut-2-ynoate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4,4,4-trifluorobut-2-ynoate

A833241-250MG A833241-1G A833241-5G
103841-176EA 66 USD
103841-176 76807-640 76845-412 76845-414
Ethyl-4,4,4-trifluorobut-2-ynoate 97%
Ethyl-4,4,4-trifluorobut-2-ynoate

Precaución: for Research Use Only

Formula: C₆H₅F₃O₂
MW: 166.099 g/mol
Boiling Pt: 116.5 °C (760 mm Hg)
Flash Pt: 24.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192167
CAS Number: 79424-03-6
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR