Ethyl-4-(benzyloxy)-3-oxobutanoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-(benzyloxy)-3-oxobutanoate

A145796-10G A145796-1G A145796-25G A145796-5G
103803-594EA 6 USD
103803-594 103841-674 76823-648 77316-976 77316-978 77316-980
Ethyl-4-(benzyloxy)-3-oxobutanoate 95%
Ethyl-4-(benzyloxy)-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19388798
CAS Number: 67354-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR