Ethyl 4-(benzyloxy)-3-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A101986-250MG A101986-1G
103796-238EA 86 USD
103796-238 77008-358 77308-816
Ethyl 4-(benzyloxy)-3-hydroxybenzoate 97%
Ethyl 4-(benzyloxy)-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
Storage Temperature: Refrigerator
MDL Number: MFCD26385855
CAS Number: 177429-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR