4-(Ethoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enecarboxylate

A306302-100MG A306302-250MG A306302-10G
103814-458EA 25 USD
103814-458 103835-626 77032-528 77032-530 77872-420
4-(Ethoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%
4-(Ethoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₅BO₄
Boiling Pt: 312.6±52.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11520546
CAS Number: 1049004-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR