Ethyl 3-oxopiperidine-4-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A509822-1G A509822-5G A509822-100G A509822-10G A509822-250MG
103828-376EA 21 USD
103828-376 103839-476 76843-462 76843-464 77034-564 77034-566 77034-568
Ethyl 3-oxopiperidine-4-carboxylate hydrochloride 95%
Ethyl 3-oxopiperidine-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄ClNO₃
MW: 207.66 g/mol
Boiling Pt: 268.5 °C (760 mm Hg)
Flash Pt: 116.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12828687
CAS Number: 72738-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR