Ethyl propionylacetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-oxopentanoate, Ethyl 3-oxovalerate

A110114-100G A110114-10G A110114-25G A110114-500G A110114-1G A110114-5G
103797-618EA 8 USD
103797-618 103823-426 76796-762 76796-764 76796-766 76796-768 77283-940 77310-226
Ethyl propionylacetate 95%
Ethyl propionylacetate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 193 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009317
CAS Number: 4949-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR