Ethyl-3-oxocyclohexanecarboxylate 96%

Supplier: Ambeed
Warning

A128840-10G A128840-250MG A128840-1G A128840-5G
103801-188EA 34 USD
103801-188 76797-788 76797-790 77313-542 77313-544
Ethyl-3-oxocyclohexanecarboxylate 96%

Precaución: for Research Use Only

Formula: C₉H₁₄O₃
Boiling Pt: 248.8 °C (760 mm Hg)
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00205583
CAS Number: 33668-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR