Ethyl-3-oxo-4-phenylbutanoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-oxo-4-phenylbutanoate

A145792-100MG A145792-250MG A145792-1G A145792-25G
103803-588EA 9 USD
103803-588 103841-668 77285-928 77285-930 77316-970 77316-972
Ethyl-3-oxo-4-phenylbutanoate 95%
Ethyl-3-oxo-4-phenylbutanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.2378 g/mol
Boiling Pt: 290.3 °C (760 mm Hg)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD03844818
CAS Number: 718-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR