Ethyl benzoylacetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

A132007-100G A132007-10G A132007-500G A132007-1000G A132007-25G
103808-434EA 165 USD
103808-434 76797-934 76797-936 76797-938 77314-220 77314-222
Ethyl benzoylacetate 95%
Ethyl benzoylacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 268.1 °C (760 mm Hg)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00009196
CAS Number: 94-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR