Ethyl picolinoylacetate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-oxo-3-(2-pyridyl)propionate, Ethyl 3-oxo-3-(pyridin-2-yl)propanoate

A464737-100G A464737-10G A464737-250MG A464737-1G A464737-25G A464737-5G
103827-966EA 8 USD
103827-966 103839-044 76804-706 76804-708 76804-710 76842-946 76842-948 76842-950
Ethyl picolinoylacetate 96%
Ethyl picolinoylacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Boiling Pt: 276.6 °C (760 mm Hg)
Flash Pt: 121.074 °C
Storage Temperature: Ambient
MDL Number: MFCD00094022
CAS Number: 26510-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR