Ethyl 3-oxo-3-(p-tolyl)propanoate 95%

Supplier: Ambeed
Warning

A535384-10G A535384-250MG A535384-1G A535384-5G
103828-590EA 21 USD
103828-590 103839-712 76726-328 76726-330 76843-732 76843-734
Ethyl 3-oxo-3-(p-tolyl)propanoate 95%
Ethyl 3-oxo-3-(p-tolyl)propanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Boiling Pt: 244…245 °C at N/A mmHg
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03424815
CAS Number: 27835-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR