Ethyl-3-methyl-1H-pyrrole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-methyl-1H-pyrrole-2-carboxylate

A124541-10G A124541-250MG A124541-25G A124541-5G A124541-1G
103800-544EA 22 USD
103800-544 77026-694 77026-696 77026-698 77297-998 77312-680
Ethyl-3-methyl-1H-pyrrole-2-carboxylate 98%
Ethyl-3-methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.1784 g/mol
Boiling Pt: 248.5±20.0 °C (760 mm Hg)
Melting Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01006747
CAS Number: 3284-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR