Ethyl-3-methyl-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-methyl-1H-pyrazole-4-carboxylate, 4-(Ethoxycarbonyl)-3-methyl-1H-pyrazole

A135601-100G A135601-10G A135601-250MG A135601-1G A135601-25G A135601-5G
103802-182EA 7 USD
103802-182 76798-028 76798-030 76798-032 77315-026 77315-028 77315-030
Ethyl-3-methyl-1H-pyrazole-4-carboxylate 97%
Ethyl-3-methyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
Melting Pt: 51…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233457
CAS Number: 85290-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR