Ethyl-3-iodopropanoate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-iodopropanoate

A110766-10G A110766-250MG A110766-25G A110766-1G A110766-5G
103797-726EA 12 USD
103797-726 103823-534 76796-808 76796-810 76796-812 77310-332 77310-334
Ethyl-3-iodopropanoate 96%
Ethyl-3-iodopropanoate

Precaución: for Research Use Only

Formula: C₅H₉IO₂
Boiling Pt: 203 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00045323
CAS Number: 6414-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR