Ethyl 3-Hydroxypropanoate 95%

Supplier: Ambeed
Warning

A190136-100G A190136-10G A190136-250MG A190136-1G A190136-25G A190136-5G
103816-262EA 31 USD
103816-262 103846-858 76998-940 76998-942 76998-944 77302-206 77302-208 77302-210
Ethyl 3-Hydroxypropanoate 95%
Ethyl 3-Hydroxypropanoate

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
Boiling Pt: 187.5 °C (760 mm Hg)
Flash Pt: 79.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11652203
CAS Number: 623-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR