Ethyl-3-hydroxyhexanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-hydroxyhexanoate

A371029-100G A371029-25G A371029-5G
103837-488EA 20 USD
103837-488 76803-590 76840-734 76840-736
Ethyl-3-hydroxyhexanoate 98%
Ethyl 3-hydroxyhexanoate

Precaución: for Research Use Only

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 241.7 °C (760 mm Hg)
Flash Pt: 94.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00036604
CAS Number: 2305-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR