Ethyl-3-hydroxycyclobutanecarboxylate 95% (mixture of isomers)
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-hydroxycyclobutanecarboxylate
A161182-100MG
A161182-250MG
A161182-1G
A161182-10G
103805-462EA
11
USD
103805-462
103843-700
76973-602
76973-604
77319-666
77611-562
Ethyl-3-hydroxycyclobutanecarboxylate 95% (mixture of isomers)
Ethyl-3-hydroxycyclobutanecarboxylate
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol Boiling Pt: 209 °C (760 mm Hg) Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD09758972 CAS Number: 17205-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...