Ethyl-3-fluoro-4-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-fluoro-4-nitrobenzoate

A250690-10G A250690-1G
103811-942EA 9 USD
103811-942 103851-780 77031-516 77091-138
Ethyl-3-fluoro-4-nitrobenzoate 98%
Ethyl-3-fluoro-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈FNO₄
MW: 213.1600 g/mol
Flash Pt: 151.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07782042
CAS Number: 914347-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR