Ethyl 3-fluoro-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A252345-1G A252345-5G
103812-036EA 27 USD
103812-036 103851-880 77304-578 77304-580
Ethyl 3-fluoro-2-methylbenzoate 98%
Ethyl 3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204676
CAS Number: 114312-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR