Ethyl 3-cyclopentyl-3-oxopropionate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-cyclopentyl-3-oxopropanoate

A162189-10G A162189-250MG A162189-1G A162189-5G
103805-586EA 41 USD
103805-586 103843-834 77273-358 77273-360 77300-574 77319-874
Ethyl 3-cyclopentyl-3-oxopropionate 97%
Ethyl 3-cyclopentyl-3-oxopropionate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₃
MW: 184.232 g/mol
Boiling Pt: 255.3 °C (760 mm Hg)
Flash Pt: 106.1 °C
Storage Temperature: Ambient
MDL Number: MFCD06410650
CAS Number: 24922-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR