Ethyl 3-Bromopyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromopicolinate

A142291-10G A142291-250MG A142291-25G A142291-1G A142291-5G
103803-138EA 16 USD
103803-138 76984-238 76984-240 76984-242 77299-284 77299-286
Ethyl 3-Bromopyridine-2-carboxylate 97%
Ethyl 3-Bromopyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
Boiling Pt: 282.4±20.0 °C (760 mm Hg)
Flash Pt: 124.6 °C
Storage Temperature: Ambient
MDL Number: MFCD13189685
CAS Number: 434319-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR