Ethyl-3-bromoisoxazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromoisoxazole-5-carboxylate

A157132-10G A157132-250MG A157132-1G A157132-5G
103804-970EA 13 USD
103804-970 103843-170 77011-034 77011-036 77300-234 77318-990
Ethyl-3-bromoisoxazole-5-carboxylate 98%
Ethyl-3-bromoisoxazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₃
MW: 220.0200 g/mol
Boiling Pt: 279.1 °C (760 mm Hg)
Melting Pt: 47…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04035587
CAS Number: 105174-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR