Ethyl-3-bromo-4-hydroxybenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-bromo-4-hydroxybenzoate
A100338-100MG
A100338-5G
103795-372EA
25
USD
103795-372
103821-074
76994-436
77282-478
Ethyl-3-bromo-4-hydroxybenzoate 97%
Ethyl-3-bromo-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₃ Boiling Pt: 297.9±20.0 °C (760 mm Hg) Melting Pt: 102.2…103.1 °C Flash Pt: 134 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06204654 CAS Number: 37470-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...