Ethyl-3-bromo-4-cyanobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromo-4-cyanobenzoate

A219318-10G A219318-250MG A219318-5G
103818-780EA 57 USD
103818-780 103849-656 76975-792 76975-794 77088-208
Ethyl-3-bromo-4-cyanobenzoate 95%
Ethyl-3-bromo-4-cyanobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13184275
CAS Number: 362527-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR