Ethyl-3-bromo-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromo-1H-pyrazole-4-carboxylate

A224524-100MG A224524-10G A224524-250MG A224524-1G A224524-5G
103819-116EA 43 USD
103819-116 103850-008 76826-268 76826-270 76826-272 77088-666 77088-668
Ethyl-3-bromo-1H-pyrazole-4-carboxylate 97%
Ethyl-3-bromo-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032195
CAS Number: 1353100-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR