Ethyl-3-bromo-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-bromoindole-2-carboxylate, Ethyl 3-Bromoindole-2-carboxylate, 3-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 3-bromo-1H-indole-2-carboxylate

A261806-250MG A261806-1G A261806-25G A261806-5G A261806-10G
103812-540EA 12 USD
103812-540 103852-430 76771-890 77092-036 77304-830 77304-832 77626-368
Ethyl-3-bromo-1H-indole-2-carboxylate 97%
Ethyl-3-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 387.9 °C (760 mm Hg)
Melting Pt: 149…151 °C
Flash Pt: 188.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02071793
CAS Number: 91348-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR