Ethyl 3-bromo-1,2,4-oxadiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A147416-100MG A147416-10G A147416-250MG
103803-774EA 88 USD
103803-774 103841-876 77272-844 77272-846 77272-848
Ethyl 3-bromo-1,2,4-oxadiazole-5-carboxylate 97%
Ethyl 3-bromo-1,2,4-oxadiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06795880
CAS Number: 121562-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR