Ethyl-3-aminopicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-aminopicolinate, 3-Aminopyridine-2-carboxylic acid ethyl ester
A119203-100G
A119203-10G
A119203-250MG
A119203-25G
A119203-1G
A119203-5G
103799-178EA
14
USD
103799-178
77271-394
77271-396
77271-398
77271-400
77311-852
77311-854
Ethyl-3-aminopicolinate 98%
Ethyl 3-aminopicolinate
Precaución: for Research Use Only
Formula:
C₈H₁₀N₂O₂ MW: 166.1800 g/mol Boiling Pt: 326.2 °C (760 mm Hg) Flash Pt: 151.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00661294 CAS Number: 27507-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...