Tricaine methanesulfonate 98%

Supplier: Ambeed
Warning

A256653-100G A256653-100MG A256653-10G A256653-500G A256653-25G A256653-5G A256653-50MG
103852-142EA 12 USD
103852-142 76827-068 76827-070 76827-072 76827-074 77304-686 77304-688 77683-781
Tricaine methanesulfonate 98%
Tricaine methanesulphonate

Precaución: For research use only.

Formula: C₁₀H₁₅NO₅S
Boiling Pt: 294.7 °C (760 mm Hg)
Melting Pt: 146…149 °C
Flash Pt: 151.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00013176
CAS Number: 886-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR