Ethyl-3-aminobenzoate 98%

Supplier: Ambeed
Warning

A144280-100G A144280-10G A144280-1G A144280-25G A144280-5G
103841-462EA 9 USD
103841-462 77247-076 77247-078 77247-080 77316-704 77316-706
Ethyl-3-aminobenzoate 98%
Ethyl-3-aminobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
Boiling Pt: 294.7 °C (760 mm Hg)
Melting Pt: 27…30 °C
Flash Pt: 151.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00007794
CAS Number: 582-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR