Ethyl-3-amino-4-pyridinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-amino-4-pyridinecarboxylate, Ethyl 3-Aminoisonicotinate, 3-Aminoisonicotinic acid ethyl ester, Ethyl 3-Aminopyridine-4-carboxylate, 3-Aminopyridine-4-carboxylic acid ethyl ester

A360871-1G A360871-25G A360871-5G A360871-10G
103826-272EA 17 USD
103826-272 103837-284 76840-496 76840-498 76840-500 77015-876
Ethyl-3-amino-4-pyridinecarboxylate 98%
Ethyl-3-amino-4-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
Boiling Pt: 303.9 °C (760 mm Hg)
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD03788270
CAS Number: 14208-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR