Ethyl-3-amino-3-methylbutanoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-amino-3-methylbutanoate hydrochloride

A103208-100G A103208-10G A103208-250MG A103208-25G A103208-1G A103208-5G
103796-506EA 7 USD
103796-506 103822-208 76970-266 76970-268 76970-270 76970-272 77309-088 77309-090
Ethyl-3-amino-3-methylbutanoate hydrochloride 95%
Ethyl-3-amino-3-methylbutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
MW: 181.6604 g/mol
Flash Pt: 95.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11844787
CAS Number: 85532-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR