Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-amino-1H-pyrrole-2-carboxylate hydrochloride

A100828-100G A100828-10G A100828-250MG A100828-25G A100828-1G A100828-5G
103795-644EA 11 USD
103795-644 103821-340 76994-482 76994-484 76994-486 76994-488 77296-850 77296-852
Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride 97%
Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O₂
MW: 190.6274 g/mol
Flash Pt: 141.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038917
CAS Number: 252932-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR