Ethyl-3,4-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate

A278162-100G A278162-10G A278162-500G A278162-5G A278162-1000G
103853-340EA 5 USD
103853-340 76692-978 76692-980 76692-982 77305-294 77626-434
Ethyl-3,4-dihydroxybenzoate 98%
Ethyl-3,4-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 358.1 °C (760 mm Hg)
Melting Pt: 132…134 °C
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00002199
CAS Number: 3943-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR