Ethyl-3,3-diethoxypropionate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,3-diethoxypropanoate, Ethyl 3,3-diethoxypropionate

A210571-25G A210571-100G A210571-10G
103848-714EA 5 USD
103848-714 77087-094 77152-416 77152-418 77303-130
Ethyl-3,3-diethoxypropionate 97%
Ethyl-3,3-diethoxypropionate

Precaución: for Research Use Only

Formula: C₉H₁₈O₄
Boiling Pt: 209.3 °C (760 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Freezer
MDL Number: MFCD00009865
CAS Number: 10601-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR