Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-(Trifluoromethyl)pyrazole-5-carboxylate
A517216-100MG
A517216-250MG
A517216-25G
A517216-1G
A517216-5G
103828-470EA
35
USD
103828-470
77017-374
77017-376
77017-378
77307-688
77613-628
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate 97%
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₇F₃N₂O₂ MW: 208.14 g/mol Boiling Pt: 288.5 °C (760 mm Hg) Melting Pt: 95 °C at N/A mmHg Flash Pt: 128.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD04111238 CAS Number: 129768-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...