Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester, Ethyl 3-trifluoromethylpyrazole-4-carboxylate
A108874-250MG
A108874-25G
A108874-1G
A108874-5G
103797-396EA
21
USD
103797-396
103823-186
76796-688
76796-690
77310-012
77310-014
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₇F₃N₂O₂ MW: 208.14 g/mol Melting Pt: 144…148 °C Flash Pt: 136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052083 CAS Number: 155377-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...