Ethyl 3-(4-iodophenyl)-3-oxopropanoate 97%

Supplier: Ambeed
Warning

A118350-250MG A118350-5G
103799-040EA 57 USD
103799-040 76797-230 77297-594
Ethyl 3-(4-iodophenyl)-3-oxopropanoate 97%
Ethyl 3-(4-iodophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁IO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03701540
CAS Number: 63131-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR