Ethyl 3-(4-fluorophenyl)propanoate 96%

Supplier: Ambeed
Warning

A223871-250MG A223871-1G
103819-072EA 8 USD
103819-072 103849-954 76814-970 77303-676
Ethyl 3-(4-fluorophenyl)propanoate 96%
Ethyl 3-(4-fluorophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
MW: 196.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772966
CAS Number: 7116-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR