Ethyl 3-(4-Cyanophenyl)-3-oxopropionate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-(4-cyanophenyl)-3-oxopropanoate, 3-(4-Cyanophenyl)-3-oxopropionic acid Ethyl ester
A206764-100MG
A206764-250MG
A206764-5G
103817-730EA
55
USD
103817-730
103848-406
77275-006
77275-008
77303-006
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate 97%
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO₃ MW: 217.22 g/mol Boiling Pt: 358.1 °C (760 mm Hg) Flash Pt: 156.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00029621 CAS Number: 49744-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...