Ethyl (4-bromobenzoyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: ethyl 3-(4-bromophenyl)-3-oxopropanoate, 3-(4-bromophenyl)-3-oxo-propionic acid ethyl ester, Benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester

A237271-10G A237271-1G A237271-25G A237271-5G A237271-100G
103811-142EA 11 USD
103811-142 103850-920 76771-320 77089-994 77089-996 77089-998 77626-262
Ethyl (4-bromobenzoyl)acetate 98%
Ethyl (4-bromobenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₃
MW: 271.11 g/mol
Boiling Pt: 332.4 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00231243
CAS Number: 26510-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR