Ethyl-3-(3-nitrophenyl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(3-nitrophenyl)-3-oxopropanoate

A100761-250MG A100761-1G A100761-5G
103795-614EA 17 USD
103795-614 103821-310 76796-244 77282-558 77282-560
Ethyl-3-(3-nitrophenyl)-3-oxopropanoate 95%
Ethyl-3-(3-nitrophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₅
MW: 237.21 g/mol
Boiling Pt: 314.4 °C (760 mm Hg)
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00126483
CAS Number: 52119-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR