Ethyl (3-fluorobenzoyl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(3-fluorophenyl)-3-oxopropanoate

A407093-1G A407093-5G A407093-100MG A407093-250MG
103827-150EA 23 USD
103827-150 103838-190 76841-682 76841-684 77168-356 77168-358
Ethyl (3-fluorobenzoyl)acetate 97%
Ethyl (3-fluorobenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
Boiling Pt: 275.1 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03424809
CAS Number: 33166-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR