Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(3-amino-4-(methylamino)-N-(pyridin-2-yl)benzamido)propanoate, 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester

A171906-100G A171906-10G A171906-25G A171906-5G
103806-676EA 6 USD
103806-676 103844-958 76784-256 76784-258 77082-132 77082-134
Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate 98%
Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate

Precaución: for Research Use Only

Formula: C₁₈H₂₂N₄O₃
MW: 342.40 g/mol
Boiling Pt: 576.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09833624
CAS Number: 212322-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR